Learn More
3-Methylglutaric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methylglutaric acid |
| CAS | 626-51-7 |
| Molecular Formula | C6 H10 O4 |
| Quantity | 10 g |
| Synonym | Pentanedioic acid','3-methyl- (9CI','ACI), 3-Methylpentanedioic acid (ACI), Glutaric acid','3-methyl- (7CI','8CI), Glutaric acid','β-methyl- (4CI), 3-Methylglutaric acid, NSC 14870, β-Methylglutaric acid |
| InChI Formula | InChI=1S/C6H10O4/c1-4(2-5(7)8)3-6(9)10/h4H,2-3H2,1H3,(H,7,8)(H,9,10) |
| SMILES | CC(CC(=O)O)CC(=O)O |
| IUPAC Name | 3-methylpentanedioic acid |
| Molecular Weight (g/mol) | 146.14 |
| Formula Weight | 146.0579 |