Learn More
3-Methylglutaric acid, MedChemExpress
Description
3-Methylglutaric acid, a leucine metabolite, is a conspicuous C6 dicarboxylic organic acid classically associated with two distinct leucine pathway enzyme deficiencies, 3-hydroxy-3-methylglutaryl CoA lyase (HMGCL) and 3-methylglutaconyl CoA hydratase (AUH).
Specifications
Specifications
| Chemical Name or Material | 3-Methylglutaric acid |
| CAS | 626-51-7 |
| Color | Colorless |
| Purity Grade Notes | Research |
| Molecular Formula | C6H10O4 |
| Solubility Information | DMSO : 100 mg/mL (684.28 mM; Need ultrasonic) |
| SMILES | O=C(O)CC(C)CC(O)=O |
| Molecular Weight (g/mol) | 146.14 |
| Quantity | 1 g |
| Formula Weight | 146.14 |
| Show More |