Learn More
3-Methylbenzenesulfonic Acid Monohydrate, TRC
Product Code. 30480835
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480835 1 g 1g
30484239 100 mg 100mg
30470666 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30480835 Supplier Toronto Research Chemicals Supplier No. TRCM2765951G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 312619-56-0
Molecular Formula C7 H8 O3 S . H2 O
Molecular Weight (g/mol) 190.22
Synonym Benzenesulfonic acid, 3-methyl-, hydrate (1:1), Benzenesulfonic acid, 3-methyl-, hydrate (1:1) (ACI), Benzenesulfonic acid, 3-methyl-, monohydrate (9CI), m-Toluenesulfonic Acid Monohydrate, GW845032A, 3-Methylbenzenesulfonic acid hydrate, m-TSA, GSK3985771C (Niraparib related)
IUPAC Name 3-methylbenzenesulfonic acid;hydrate
SMILES O.Cc1cccc(c1)S(=O)(=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C7H8O3S.H2O/c1-6-3-2-4-7(5-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2
Formula Weight 190.023
Chemical Name or Material 3-Methylbenzenesulfonic Acid Monohydrate