Learn More
3-Methyl-5-oxo-5-phenylvaleric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-5-oxo-5-phenylvaleric Acid |
| CAS | 2840-61-1 |
| Molecular Formula | C12H14O3 |
| Quantity | 1 g |
| Synonym | beta-Methyl-delta-oxo-benzenepentanoic Acid, 4-Benzoyl-3-methyl-butyric Acid |
| InChI Formula | InChI=1S/C12H14O3/c1-9(8-12(14)15)7-11(13)10-5-3-2-4-6-10/h2-6,9H,7-8H2,1H3,(H,14,15) |
| SMILES | CC(CC(=O)O)CC(=O)c1ccccc1 |
| IUPAC Name | 3-methyl-5-oxo-5-phenylpentanoic acid |
| Molecular Weight (g/mol) | 206.24 |
| Formula Weight | 206.0943 |