missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzamide, 97%, Thermo Scientific™

Product Code. p-7027715
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11480568

Brand: Thermo Scientific Alfa Aesar B24691.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Methyl-4-nitrobenzamide is used as synthetic organic intermediates.

Solubility
Soluble in alcohol, slightly soluble in hot water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99584-85-7
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.16
MDL Number MFCD00031468
InChI Key BCAAGQMIPKPDDX-UHFFFAOYSA-N
Synonym 3-methyl-4-nitro-benzamide, 3-methyl-4-nitro benzamide, #, benzamide, 3-methyl-4-nitro, 3-methyl-4-nitrobenzoic acid amide
PubChem CID 595900
IUPAC Name 3-methyl-4-nitrobenzamide
SMILES CC1=CC(=CC=C1[N+]([O-])=O)C(N)=O

Specifications

Melting Point 158°C to 160°C
Quantity 1 g
Beilstein 2264048
Solubility Information Soluble in alcohol,slightly soluble in hot water.
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.