Learn More
3-[Methyl(3-methylbutyl)amino]propanenitrile, TRC
Spezifikation
Spezifikation
| Chemical Name or Material | 3-[Methyl(3-methylbutyl)amino]propanenitrile |
| CAS | 1019125-05-3 |
| Molecular Formula | C9H18N2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C9H18N2/c1-9(2)5-8-11(3)7-4-6-10/h9H,4-5,7-8H2,1-3H3 |
| SMILES | CC(C)CCN(C)CCC#N |
| IUPAC Name | 3-[methyl(3-methylbutyl)amino]propanenitrile |
| Molecular Weight (g/mol) | 154.25 |
| Formula Weight | 154.147 |