Learn More
3-Methoxy-7Beta-methyl-estra-2,5(10)-dien-17-one, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methoxy-7beta-methyl-estra-2,5(10)-dien-17-one |
| CAS | 32297-43-1 |
| Molecular Formula | C20H28O2 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C20H28O2/c1-12-10-13-11-14(22-3)4-5-15(13)16-8-9-20(2)17(19(12)16)6-7-18(20)21/h4,12,16-17,19H,5-11H2,1-3H3/t12-,16+,17-,19+,20-/m0/s1 |
| SMILES | COC1=CCC2=C(C[C@H](C)[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CCC4=O)C1 |
| IUPAC Name | (7S,8R,9S,13S,14S)-3-methoxy-7,13-dimethyl-4,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-one |
| Molecular Weight (g/mol) | 300.44 |
| Formula Weight | 300.2089 |