missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxy-5-(trifluoromethyl)cinnamic acid, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Methoxy-5-(trifluoromethyl)cinnamic acid, is used as a pharmaceutical intermediate.
Solubility
Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids and strong bases.
3-Methoxy-5-(trifluoromethyl)cinnamic acid, is used as a pharmaceutical intermediate.
Solubility
Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids and strong bases.
Specifications
Specifications
| Melting Point | 155°C to 157°C |
| Quantity | 250 mg |
| Solubility Information | Soluble in organic solvents. |
| Formula Weight | 246.18 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Methoxy-5-(trifluoromethyl)cinnamic acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?