Learn More
3-Methoxy-2-nitrobenzoic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Methoxy-2-nitrobenzoic acid |
| CAS | 4920-80-3 |
| Molecular Formula | C8H7NO5 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C8H7NO5/c1-14-6-4-2-3-5(8(10)11)7(6)9(12)13/h2-4H,1H3,(H,10,11) |
| SMILES | COc1cccc(C(=O)O)c1[N+](=O)[O-] |
| IUPAC Name | 3-methoxy-2-nitrobenzoic acid |
| Molecular Weight (g/mol) | 197.15 |
| Formula Weight | 197.0324 |