Learn More
3-Mercaptopicolinic Acid Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Mercaptopicolinic Acid, Hydrochloride |
| CAS | 320386-54-7 |
| Molecular Formula | C6 H5 N O2 S . Cl H |
| Quantity | 5 g |
| Synonym | 3-Mercapto-2-pyridinecarboxylic acid hydrochloride |
| InChI Formula | InChI=1S/C6H5NO2S.ClH/c8-6(9)5-4(10)2-1-3-7-5;/h1-3,10H,(H,8,9);1H |
| SMILES | Cl.OC(=O)c1ncccc1S |
| IUPAC Name | 3-sulfanylpyridine-2-carboxylic acid;hydrochloride |
| Molecular Weight (g/mol) | 191.64 |
| Formula Weight | 190.9808 |