missing translation for 'onlineSavingsMsg'
Learn More

3-Isoniazid, TRC

Product Code. 17901345
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901345 1 g 1g
30137804 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17901345 Supplier Toronto Research Chemicals Supplier No. TRCI8214601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-53-7
InChI Formula InChI=1S/C6H7N3O/c7-9-6(10)5-2-1-3-8-4-5/h1-4H,7H2,(H,9,10)
IUPAC Name pyridine-3-carbohydrazide
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
Chemical Name or Material 3-Isoniazid

Specifications

Quantity 1 g
Synonym Nicotinic Acid Hydrazide, Nicotinohydrazide, 3-Pyridinecarboxylic Acid Hydrazide, (3-Pyridylcarbonyl)hydrazine, 3-Pyridinecarbohydrazide, 3-Pyridinecarboxylic Hydrazide, 3-Pyridoylhydrazine;
SMILES C(C=1C=CC=NC1)(=O)NN.Cl.Cl
Formula Weight 137.0589
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.