Learn More
3-Iodoanisole, TRC
Product Code. 30399183
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30399183 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30399183 Supplier Toronto Research Chemicals Supplier No. TRCI6868502.5G
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Iodoanisole
CAS 766-85-8
Molecular Formula C7H7IO
Quantity 2.5 g
Synonym 1-Iodo-3-methoxy-benzene, 1-Iodo-3-methoxybenzene, 3-Methoxyphenyl Iodide, m-Methoxyiodobenzene, NSC 60726
InChI Formula InChI=1S/C7H7IO/c1-9-7-4-2-3-6(8)5-7/h2-5H,1H3
SMILES COc1cccc(I)c1
IUPAC Name 1-iodo-3-methoxybenzene
Molecular Weight (g/mol) 234.03
Formula Weight 233.9542