Learn More
3-Indoxyl Sulfate-d5 Potassium (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 3-Indoxyl Sulfate-d5 Potassium Salt |
| CAS | 1644451-34-2 |
| Molecular Formula | C8HD5KNO4S |
| Quantity | 10 mg |
| Synonym | 1H-Indol-3-ol 3-(Hydrogen Sulfate)-d5 Potassium Salt, 1H-Indol-3-ol Hydrogen Sulfate (Ester)-d5 Monopotassium Salt, Indol-3-yl Potassium Sulfate-d5, Indol-3-ol Potassium Sulfate-d5, Indol-3-yl Sulfate Potassium Salt-d5, Potassium Indol-3-yl Sulfate-d5 |
| InChI Formula | InChI=1S/C8H7NO4S.K/c10-14(11,12)13-8-5-9-7-4-2-1-3-6(7)8;/h1-5,9H,(H,10,11,12);/q;+1/p-1/i1D,2D,3D,4D,5D; |
| SMILES | [2H]C1=C2C(C(OS(=O)(O[K])=O)=C([2H])N2)=C([2H])C([2H])=C1[2H] |
| Molecular Weight (g/mol) | 256.33 |