Learn More
3-Hydroxy Tolperisone Maleate, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy Tolperisone Maleate |
| CAS | 283585-02-4 |
| Molecular Formula | C20H27NO6 |
| Quantity | 100 mg |
| Synonym | 1-(3-Hydroxy-4-methylphenyl)-2-methyl-3-(1-piperidinyl)-1-propanone (2Z)-2-Butenedioate |
| InChI Formula | InChI=1S/C16H23NO2.C4H4O4/c1-12-6-7-14(10-15(12)18)16(19)13(2)11-17-8-4-3-5-9-17;5-3(6)1-2-4(7)8/h6-7,10,13,18H,3-5,8-9,11H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CC1=C(C=C(C=C1)C(=O)C(C)CN2CCCCC2)O.C(=C\C(=O)O)\C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;1-(3-hydroxy-4-methylphenyl)-2-methyl-3-piperidin-1-ylpropan-1-one |
| Molecular Weight (g/mol) | 377.43 |
| Formula Weight | 377.1838 |