Learn More
3-Hydroxy Nevirapine-d3, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy Nevirapine-d3 |
| CAS | 1346602-85-4 |
| Molecular Formula | C15H11D3N4O2 |
| Quantity | 0.5 mg |
| Synonym | 11-Cyclopropyl-5,11-dihydro-3-hydroxy-4-methy-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one-d3 |
| InChI Formula | InChI=1S/C15H14N4O2/c1-8-11(20)7-17-14-12(8)18-15(21)10-3-2-6-16-13(10)19(14)9-4-5-9/h2-3,6-7,9,20H,4-5H2,1H3,(H,18,21)/i1D3 |
| SMILES | [2H]C([2H])([2H])c1c(O)cnc2N(C3CC3)c4ncccc4C(=O)Nc12 |
| IUPAC Name | 11-cyclopropyl-3-hydroxy-4-(trideuteriomethyl)-5H-dipyrido[2,3-d:3',2'-f][1,4]diazepin-6-one |
| Molecular Weight (g/mol) | 285.32 |
| Formula Weight | 285.1305 |