missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxybenzyl alcohol, 98%, Thermo Scientific™

Product Code. p-7022154
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11492507

Brand: Thermo Scientific Alfa Aesar A14786.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4383-06-6
MDL Number MFCD00004644

Specifications

Melting Point 135°C to 137°C
Molecular Formula C8H10O3
Quantity 10 g
Beilstein 1867860
Synonym 3-hydroxy-4-methoxybenzyl alcohol, 5-hydroxymethyl-2-methoxyphenol, isovanillyl alcohol, 3-hydroxy-4-methoxybenzylalcohol, benzenemethanol, 3-hydroxy-4-methoxy, 4-methoxy-3-hydroxy benzyl alcohol, pubchem15050, ksc495i5l, 3-hydroxy-4-methoxy-benzyl alcohol, 3-hydroxy-4-methoxy benzyl alcohol
InChI Key WHKRHBLAJFYZKF-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)CO)O
IUPAC Name 5-(hydroxymethyl)-2-methoxyphenol
Molecular Weight (g/mol) 154.17
PubChem CID 78089
Formula Weight 154.17
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-4-methoxybenzyl alcohol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.