Learn More
3-Hydroxy-1-methyl-pyridinium Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy-1-methylpyridin-1-ium bromide |
| CAS | 31034-86-3 |
| Molecular Formula | C6H8NO.Br |
| Quantity | 25 g |
| Synonym | 3-Hydroxy-1-methylpyridinium Bromide, Pyridinium, 3-hydroxy-1-methyl-, bromide (1:1), 3-Hydroxy-1-methylpyridinium bromide (6CI), Pyridinium, 3-hydroxy-1-methyl-, bromide (8CI,9CI), 3-Hydroxy-N-methylpyridinium bromide, N-Methyl-3-hydroxypyridinium bromide, Ro 1-5237 |
| InChI Formula | InChI=1S/C6H7NO.BrH/c1-7-4-2-3-6(8)5-7;/h2-5H,1H3;1H |
| SMILES | [Br-].C[n+]1cccc(O)c1 |
| IUPAC Name | 1-methylpyridin-1-ium-3-ol;bromide |
| Molecular Weight (g/mol) | 190.04 |
| Formula Weight | 188.979 |