Learn More
3-(Formylamino)-1H-pyrazole-4-carboxylic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 5-(Formylamino)-1H-pyrazole-4-carboxylate |
| CAS | 31055-19-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 H9 N3 O3 |
| Quantity | 1 g |
| Synonym | Ethyl 5-(Formylamino)-1H-pyrazole-4-carboxylate, Allopurinol Imp. E (EP) |
| InChI Formula | InChI=1S/C7H9N3O3/c1-2-13-7(12)5-3-9-10-6(5)8-4-11/h3-4H,2H2,1H3,(H2,8,9,10,11) |
| SMILES | CCOC(=O)c1cn[nH]c1NC=O |
| IUPAC Name | ethyl 5-formamido-1H-pyrazole-4-carboxylate |
| Molecular Weight (g/mol) | 183.16 |
| Show More |