Learn More
3-Fluoro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific™
Product Code. 11386689
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386689 5 g 5g
11376689 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11386689 Supplier Thermo Scientific Alfa Aesar Supplier No. H26238.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 454-67-1
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.12
MDL Number MFCD00061315
InChI Key WQKQODZOQAFYPR-UHFFFAOYSA-N
Synonym 3-amino-5-fluorobenzotrifluoride, 3-fluoro-5-trifluoromethyl aniline, 3-fluoro-5-trifluoromethyl-phenylamine, 3-fluoro-5-trifluorobenzenamine, 3-fluoro-5-trifluoromethyl-aniline, 5-fluoro-3-trifluoromethyl aniline, benzenamine, 3-fluoro-5-trifluoromethyl, 3-fluoro-5-trifluoromethyl phenylamine, pubchem1541
PubChem CID 2735913
IUPAC Name 3-fluoro-5-(trifluoromethyl)aniline
SMILES NC1=CC(F)=CC(=C1)C(F)(F)F
Specifications
Odor Characteristic
Quantity 5 g
UN Number UN2810
Formula Weight 179.12
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)aniline