missing translation for 'onlineSavingsMsg'
Learn More

3-exo-Hydroxymethylbicyclo[2.2.1]heptyl-2-exo-amine, hydrochloride, 97%, Thermo Scientific™

Product Code. 10606281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10606281 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10606281 Supplier Thermo Scientific Acros Supplier No. 265932500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 205639-91-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 174°C to 175°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H15NO·HCl
MDL Number MFCD00216898
Quantity 250 mg
Synonym diexo-3-amino-bicyclo 2.2.1 hept-2-yl-methanol hydrochloride, 1s,2r,3s,4r-3-aminobicyclo 2.2.1 heptan-2-yl methanol hydrochloride
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
InChI Key JBSSFLCCAAVUFA-SZPCUAKFSA-N
SMILES C1CC2CC1C(C2N)CO.Cl
IUPAC Name [(1S,2R,3S,4R)-3-amino-2-bicyclo[2.2.1]heptanyl]methanol;hydrochloride
Molecular Weight (g/mol) 177.67
PubChem CID 43865920
Formula Weight 177.67
Percent Purity 97%
Physical Form Crystalline Chunks and Powder
Chemical Name or Material 3-exo-Hydroxymethylbicyclo[2.2.1]heptyl-2-exo-amine, hydrochloride, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.