missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-pentanol, 97%

Product Code. p-7043844
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312006 10 g 10g
11322006 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312006 Supplier Thermo Scientific Alfa Aesar Supplier No. L03025.09
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 597-49-9 | C7H16O | 116.20 g/mol

3-Ethyl-3-pentanol was used to study the effect of alkanols on the micropolarity and microviscosity of the head group region, in the reverse micelles of AOT-heptane-water by fluorescence probing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-3-pentanol was used to study the effect of alkanols on the micropolarity and microviscosity of the head group region, in the reverse micelles of AOT-heptane-water by fluorescence probing.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store away from acids, acid chlorides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-49-9
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00004484
InChI Key XKIRHOWVQWCYBT-UHFFFAOYSA-N
Synonym 3-ethyl-3-pentanol, triethylcarbinol, 3-pentanol, 3-ethyl, triethylmethanol, triethylmethyl alcohol, tert-heptanol, 3-aethyl-pentanol-3, 3-aethyl-pentanol-3 german, 3-ethyl-3-hydroxypentane, ethyl-3 pentanol-3
PubChem CID 11702
IUPAC Name 3-ethylpentan-3-ol
SMILES CCC(O)(CC)CC

Specifications

Melting Point -13°C
Density 0.846
Boiling Point 143°C
Flash Point 40°C (104°F)
Odor Characteristic
Linear Formula (CH3CH2)3COH
Refractive Index 1.43
Quantity 10 g
UN Number UN1987
Beilstein 1731643
Solubility Information Slightly soluble in water. Soluble in alcohol.
Formula Weight 116.2
Percent Purity 97%
Chemical Name or Material 3-Ethyl-3-pentanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Ethyl-3-pentanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: 3-Ethyl-3-pentanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.