Learn More
3-Ethoxy-2,6-difluorobenzylamine, 97%, Thermo Scientific™
Product Code. 15474177
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15474177 250 mg 250mg
15464177 1 g 1g
2 options
Product Code. 15474177
Brand: Thermo Scientific Alfa Aesar H31587.MD
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1092461-29-4
Molecular Formula C9H11F2NO
Molecular Weight (g/mol) 187.19
MDL Number MFCD11519371
InChI Key GIDMHGPRTGIRRF-UHFFFAOYSA-N
Synonym 3-ethoxy-2,6-difluorobenzylamine, 1-3-ethoxy-2,6-difluorophenyl methanamine, 3-ethoxy-2,6-difluorobenzylamine, jrd, 3-ethoxy-2,6-difluorophenyl methylamine
PubChem CID 55275767
IUPAC Name (3-ethoxy-2,6-difluorophenyl)methanamine
SMILES CCOC1=C(F)C(CN)=C(F)C=C1
Specifications
Refractive Index 1.498
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 187.19
Percent Purity 97%
Chemical Name or Material 3-Ethoxy-2,6-difluorobenzylamine
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.