Learn More
3-Ethenyl Nevirapine, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Ethenyl Nevirapine |
| CAS | 284686-22-2 |
| Molecular Formula | C17H16N4O |
| Quantity | 10 mg |
| Synonym | 11-Cyclopropyl-3-ethenyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one |
| InChI Formula | InChI=1S/C17H16N4O/c1-3-11-9-19-16-14(10(11)2)20-17(22)13-5-4-8-18-15(13)21(16)12-6-7-12/h3-5,8-9,12H,1,6-7H2,2H3,(H,20,22) |
| SMILES | Cc1c(C=C)cnc2N(C3CC3)c4ncccc4C(=O)Nc12 |
| Molecular Weight (g/mol) | 292.34 |
| Formula Weight | 292.1324 |