Learn More
[3-(Dimethylamino)propyl]triphenylphosphonium Bromide Hydrobromide, TRC
Product Code. 30369752
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369752 5 g
30385963 10 g
30384109 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30369752 Supplier Toronto Research Chemicals Supplier No. TRCD4953035G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27710-82-3
Molecular Formula C23H28Br2NP
Molecular Weight (g/mol) 509.3
Synonym Phosphonium, [3-(dimethylamino)propyl]triphenyl-, bromide, hydrobromide (1:1:1) (ACI), Phosphonium, [3-(dimethylamino)propyl]triphenyl-, bromide, hydrobromide (8CI, 9CI), [3-(Dimethylamino)propyl]triphenylphosphonium bromide hydrobromide
SMILES Br.[Br-].CN(C)CCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
Specifications
Quantity 5 g
InChI Formula InChI=1S/C23H27NP.2BrH/c1-24(2)19-12-20-25(21-13-6-3-7-14-21,22-15-8-4-9-16-22)23-17-10-5-11-18-23;;/h3-11,13-18H,12,19-20H2,1-2H3;2*1H/q+1;;/p-1
Formula Weight 507.0326
Chemical Name or Material [3-(Dimethylamino)propyl]triphenylphosphonium Bromide Hydrobromide