Learn More
3-(Diethylphosphono)propanoic acid (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Diethylphosphono)propanoic acid (Technical Grade) |
| CAS | 3095-96-3 |
| Molecular Formula | C21 H28 O6 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C21H28O6/c1-19-6-5-12(23)7-11(19)3-4-13-14-8-16(25)21(27,17(26)10-22)20(14,2)9-15(24)18(13)19/h5-7,13-16,18,22,24-25,27H,3-4,8-10H2,1-2H3/t13-,14+,15-,16-,18-,19-,20-,21+/m0/s1 |
| SMILES | C[C@]12C[C@H](O)[C@@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@H]1C[C@H](O)[C@@]2(O)C(=O)CO |
| IUPAC Name | (8S,9R,10R,11S,13S,14R,16S,17R)-11,16,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 376.443 |
| Formula Weight | 376.189 |