missing translation for 'onlineSavingsMsg'
Learn More

3-(Di-tert-butylphosphino)propylamine, 10% w/w soln. in THF, Thermo Scientific™

Product Code. 15416126
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416126 5 g 5g
15406126 1 g 1g
15426126 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15416126 Supplier Thermo Scientific Alfa Aesar Supplier No. H60943.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1196147-72-4
Molecular Formula C11H26NP
Molecular Weight (g/mol) 203.31
MDL Number MFCD17013990
InChI Key PREMCOLELPAPBB-UHFFFAOYSA-N
Synonym 3-di-t-butylphosphino propylamine, 3-aminopropyldi-tert-butylphosphine, 3-di-tert-butylphosphino propylamine, 3-aminopropyl di-tert-butyl phosphane
PubChem CID 72376443
IUPAC Name 3-ditert-butylphosphanylpropan-1-amine
SMILES CC(C)(C)P(CCCN)C(C)(C)C

Specifications

Linear Formula [(CH3)3C]2PCH2CH2CH2NH2
Quantity 5 g
UN Number UN2733
Sensitivity Air Sensitive
Formula Weight 203.3
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material 3-(Di-tert-butylphosphino)propylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.