Learn More
3-Desmethyl Trimethoprim, TRC
Specifications
Specifications
| CAS | 27653-69-6 |
| Molecular Formula | C13 H16 N4 O3 |
| Quantity | 10 mg |
| Synonym | 5-[(2,4-Diamino-5-pyrimidinyl)methyl]-2,3-dimethoxy-phenol, α-(2,4-Diamino-5-pyrimidinyl)-5,6-dimethoxy-m-cresol, 2,4-Diamino-5-(3-hydroxy-4,5-dimethoxybenzyl)pyrimidine, 3-Demethyltrimethoprim, 3'-Hydroxytrimethoprim |
| InChI Formula | InChI=1S/C13H16N4O3/c1-19-10-5-7(4-9(18)12(10)20-2)3-8-6-11(14)17-13(15)16-8/h4-6,18H,3H2,1-2H3,(H4,14,15,16,17) |
| SMILES | COc1cc(Cc2cc(N)nc(N)n2)cc(O)c1OC |
| IUPAC Name | 5-[(2,6-diaminopyrimidin-4-yl)methyl]-2,3-dimethoxy-phenol |
| Molecular Weight (g/mol) | 276.29 |
| Formula Weight | 276.1222 |
| Chemical Name or Material | 3-Desmethyl Trimethoprim |