Learn More
3-Deoxy-D-erythro-hexos-2-ulose-bis-benzoylhydrazone, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Deoxy-D-erythro-hexos-2-ulose-bis-benzoylhydrazone |
| CAS | 32443-70-2 |
| Molecular Formula | C20 H22 N4 O5 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C20H22N4O5/c25-13-18(27)17(26)11-16(22-24-20(29)15-9-5-2-6-10-15)12-21-23-19(28)14-7-3-1-4-8-14/h1-10,12,17-18,25-27H,11,13H2,(H,23,28)(H,24,29)/b21-12+,22-16-/t17-,18+/m0/s1 |
| SMILES | OC[C@@H](O)[C@@H](O)C\C(=N\NC(=O)c1ccccc1)\C=N\NC(=O)c2ccccc2 |
| IUPAC Name | N-[(E)-[(2Z,4S,5R)-2-(benzoylhydrazinylidene)-4,5,6-trihydroxyhexylidene]amino]benzamide |
| Molecular Weight (g/mol) | 398.41 |
| Formula Weight | 398.159 |