missing translation for 'onlineSavingsMsg'
Learn More

3-Deazaadenosine hydrochloride, MedChemExpress

Product Code. 16430479
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16430479

Brand: Medchem Express HYW013332A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Deazaadenosine (hydrochloride) is an inhibitor of S-adenosylhomocysteine hydrolase, with a Ki of 3.9 μM; 3-Deazaadenosine has anti-inflammatory, anti-proliferative and anti-HIV activity.

3-Deazaadenosine (hydrochloride) is an inhibitor of S-adenosylhomocysteine hydrolase, with a Ki of 3.9 μM

Specifications

Chemical Name or Material 3-Deazaadenosine hydrochloride
CAS 86583-19-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H15ClN4O4
Quantity 10 mg
Solubility Information DMSO : 41.67 mg/mL (137.66 mM; Need ultrasonic)
SMILES O[C@H]1[C@@H](O[C@H](CO)[C@H]1O)N2C3=C(C(N)=NC=C3)N=C2.Cl
Molecular Weight (g/mol) 302.71
Formula Weight 302.71
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.