Learn More
3-Cyano-4-(trifluoromethyl)pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Cyano-4-(trifluoromethyl)pyridine |
| CAS | 13600-43-6 |
| Molecular Formula | C7H3F3N2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C7H3F3N2/c8-7(9,10)6-1-2-12-4-5(6)3-11/h1-2,4H |
| SMILES | FC(F)(F)C1=C(C#N)C=NC=C1 |
| IUPAC Name | 4-(trifluoromethyl)pyridine-3-carbonitrile |
| Molecular Weight (g/mol) | 172.11 |
| Formula Weight | 172.0248 |