missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-fluorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 11309905
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309905 1 g 1g
11329905 250 mg 250mg
11319905 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309905 Supplier Thermo Scientific Alfa Aesar Supplier No. L19885.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-23-1
Molecular Formula C7H3ClFNO2S
Molecular Weight (g/mol) 219.614
MDL Number MFCD03093760
InChI Key KDVZADPGGPBAAK-UHFFFAOYSA-N
Synonym 4-fluoro-3-cyanobenzenesulfonyl chloride, 3-cyano-4-fluorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-cyano-4-fluoro, 3-cyano-4-fluorobenzenesulphonyl chloride, 3-cyano-4-fluoro-benzenesulfonyl chloride, 4-fluoro-3-cyanobenzenesulfonylchloride, 4-fluoro-3-cyanobenzenesulphonyl chloride, 5-chlorosulfonyl-2-fluorobenzenecarbonitrile, pubchem5110, acmc-20a3ve
PubChem CID 2778908
IUPAC Name 3-cyano-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C#N)F

Specifications

Melting Point 67°C to 70°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 219.62
Percent Purity 98%
Chemical Name or Material 3-Cyano-4-fluorobenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.