Learn More
3-Chlorothiophene-2-boronic Acid (>80%), TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chlorothiophene-2-boronic Acid |
| CAS | 324024-80-8 |
| Molecular Formula | C4 H4 B Cl O2 S |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C4H4BClO2S/c6-3-1-2-9-4(3)5(7)8/h1-2,7-8H |
| SMILES | OB(O)c1sccc1Cl |
| IUPAC Name | (3-chlorothiophen-2-yl)boronic acid |
| Molecular Weight (g/mol) | 162.402 |
| Formula Weight | 161.971 |