missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroperoxybenzoic acid, 50-55%, cont. ca 10% 3-chlorobenzoic acid, balance water

Product Code. p-7043332
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339973 25 g 25g
11349973 100 g 100g
11359973 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339973 Supplier Thermo Scientific Alfa Aesar Supplier No. L00286.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 937-14-4 | C7H5ClO3 | 172.56 g/mol

3-Chloroperoxybenzoic acid is a strong oxidizing agent used in the oxidation reactions such as aldehydes and ketones to esters (Bayer-Villiger-Oxidation), olefines to epoxides, sulfides to sulfoxides and sulfones, and amines to nitroalkanes, N-oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloroperoxybenzoic acid is a strong oxidizing agent used in the oxidation reactions such as aldehydes and ketones to esters (Bayer-Villiger-Oxidation), olefines to epoxides, sulfides to sulfoxides and sulfones, and amines to nitroalkanes, N-oxides.

Solubility
Soluble in potassium hydroxide, chloroform and acetone. Insoluble in water.

Notes
Heat sensitive. Incompatible with reactive with reducing agents, combustible materials, organic materials and alkalies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937-14-4
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD00002127
InChI Key NHQDETIJWKXCTC-UHFFFAOYSA-N
Synonym 3-chloroperoxybenzoic acid, 3-chloroperbenzoic acid, mcpba, m-chloroperbenzoic acid, m-chloroperoxybenzoic acid, meta-chloroperoxybenzoic acid, benzenecarboperoxoic acid, 3-chloro, m-chlorobenzoyl hydroperoxide, meta-chloroperbenzoic acid, 3-chlorobenzoperoxoic acid
PubChem CID 70297
ChEBI CHEBI:52091
IUPAC Name 3-chlorobenzenecarboperoxoic acid
SMILES OOC(=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 64°C to 66°C
Quantity 25 g
UN Number UN3106
Beilstein 608317
Solubility Information Soluble in potassium hydroxide,chloroform and acetone. Insoluble in water.
Formula Weight 172.57
Percent Purity 50 to 55%
Assay cont. ≈10% 3-chlorobenzoic acid, balance water
Chemical Name or Material 3-Chloroperoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.