Learn More
3-Chlorobenzoic-d4 Acid, CDN
Product Code. 30327775
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327775 0.05 g 0.05g
30327622 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30327775 Supplier CDN Supplier No. CDND75940.05G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chlorobenzoic Acid-D4
CAS 2098655-72-0
Molecular Formula C7 D4 H Cl O2
Quantity 0.05 g
Synonym Benzoic-2,3,4,6-d4 acid, 5-chloro- (ACI), 5-Chlorobenzoic-2,3,4,6-d4 acid (ACI), 3-Chloro-2,4,5,6-tetradeuterio-benzoic acid, 3-Chlorobenzoic-d4 Acid, 3-Chlorobenzoic Acid-d4, 3-Chlorobenzoic Acid-D4
InChI Formula InChI=1 S/C7H5ClO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4 H,(H,9,10)/i1D,2 D,3 D,4 D
SMILES [2 H]c1c([2 H])c(Cl)c([2 H])c(C(=O)O)c1[2 H]
IUPAC Name 3-chloro-2,4,5,6-tetradeuteriobenzoic acid
Molecular Weight (g/mol) 160.5911
Formula Weight 160.0229 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity