missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-N,N-dimethylbenzamide, 97%, Thermo Scientific™

Product Code. 15445138
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445138 500 mg 500mg
15435138 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445138 Supplier Thermo Scientific Alfa Aesar Supplier No. H58964.ME
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24167-52-0
Molecular Formula C9H10ClNO
Molecular Weight (g/mol) 183.64
MDL Number MFCD01214035
InChI Key JSXDHOISXRKLGG-UHFFFAOYSA-N
Synonym benzamide, 3-chloro-n,n-dimethyl, n,n-dimethyl-m-chloro-benzamide, 3-cl-c6h4con ch3 2, aronis25695, 3-chlorophenyl-n,n-dimethylcarboxamide
PubChem CID 4466993
IUPAC Name 3-chloro-N,N-dimethylbenzamide
SMILES CN(C)C(=O)C1=CC(Cl)=CC=C1

Specifications

Quantity 500 mg
Formula Weight 183.64
Percent Purity 97%
Chemical Name or Material 3-Chloro-N,N-dimethylbenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.