Learn More
3-Chloro-L-alanine, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-L-alanine |
| CAS | 2731-73-9 |
| Molecular Formula | C3 H6 Cl N O2 |
| Quantity | 5 g |
| Synonym | L-Alanine, 3-chloro-, Alanine, 3-chloro-, L- (8CI), 3-Chloro-L-alanine, (2R)-β-Chloroalanine, L-3-Chloroalanine, L-β-Chloroalanine, β-Chloro-L-alanine, (2R)-2-Amino-3-chloro-propanoic acid |
| InChI Formula | InChI=1S/C3H6ClNO2/c4-1-2(5)3(6)7/h2H,1,5H2,(H,6,7)/t2-/m0/s1 |
| SMILES | N[C@@H](CCl)C(=O)O |
| IUPAC Name | (2R)-2-amino-3-chloro-propanoic acid |
| Molecular Weight (g/mol) | 123.538 |
| Formula Weight | 123.0087 |