Learn More
3-Chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Product Code. p-7031337
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15425097
Brand: Thermo Scientific Alfa Aesar H32376.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 85148-26-1
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042227
InChI Key OMRCXTBFBBWTDL-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl pyridine, 3-trifluoromethyl-5-chloropyridine, pyridine, 3-chloro-5-trifluoromethyl, pubchem3552, acmc-209zee, ksc495c1f, 3-chloro-5-trifluoromethyl-pyridine, 3-chloro-5-trifluoromethyl pyridine-carbonitrile
PubChem CID 2736698
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(Cl)=CN=C1
Specifications
Boiling Point 50°C to 51°C (4 mmHg)
Quantity 1 g
UN Number UN2810
Formula Weight 181.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)pyridine