missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-5-(trifluoromethyl)benzene-1,2-diamine, 97%, Thermo Scientific™

Product Code. 10133114
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10133114 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10133114 Supplier Thermo Scientific Maybridge Supplier No. BTB05127EA
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 132915-80-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H6ClF3N2
MDL Number MFCD00042198
Quantity 10 g
Synonym 3-chloro-5-trifluoromethyl benzene-1,2-diamine, 3-chloro-4,5-diaminobenzotrifluoride, 1,2-diamino-3-chloro-5-trifluoromethyl benzene, 3-chloro-5-trifluoromethyl-1,2-phenylenediamine, 3-chloro-5-trifluoromethyl-1,2-benzenediamine, timtec-bb sbb003446, buttpark 75\04-15, 1,2-diamino-3-chloro-5trifluoromethylbenzene
InChI Key NVNLHLRVSHJKBN-UHFFFAOYSA-N
SMILES NC1=CC(=CC(Cl)=C1N)C(F)(F)F
IUPAC Name 3-chloro-5-(trifluoromethyl)benzene-1,2-diamine
Molecular Weight (g/mol) 210.58
PubChem CID 735784
Formula Weight 210.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzene-1, 2-diamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.