missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylbenzenesulfonyl chloride, 99%, Thermo Scientific™

Product Code. p-7047885
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339905 1 g 1g
11349905 5 g 5g
11359905 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339905 Supplier Thermo Scientific Alfa Aesar Supplier No. L19886.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42413-03-6
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
MDL Number MFCD00203987
InChI Key GNYVVCRRZRVBDD-UHFFFAOYSA-N
Synonym 3-chloro-4-methylbenzene-1-sulfonyl chloride, 3-chloro-4-methylbenzenesulphonyl chloride, pubchem5098, acmc-209jpd, 2-chloro-p-toluenesulfonyl chloride, chloro 3-chloro-4-methylphenyl sulfone, 3-chloro-4-methylbenzenesulphonylchloride, 3-chloro-4-methylbenzensulfonyl chloride, 3-chloro-4-methylphenylsulfonyl chloride, 2-chloro-toluene-4-sulfonic acid chloride
PubChem CID 259744
IUPAC Name 3-chloro-4-methylbenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)S(=O)(=O)Cl)Cl

Specifications

Melting Point 34°C to 38°C
Boiling Point 155°C to 156°C (19 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Beilstein 2364094
Sensitivity Moisture sensitive
Formula Weight 225.09
Percent Purity 99%
Chemical Name or Material 3-Chloro-4-methylbenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.