Learn More
3-Chloro-2,4-dimethyl-benzenesulfonyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-2,4-dimethyl-benzenesulfonyl Chloride |
| CAS | 1783755-80-5 |
| Molecular Formula | C8H8Cl2O2S |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C8H8Cl2O2S/c1-5-3-4-7(13(10,11)12)6(2)8(5)9/h3-4H,1-2H3 |
| SMILES | Cl[S](=O)(C(C=CC(C)=C1Cl)=C1C)=O |
| IUPAC Name | 3-chloro-2,4-dimethylbenzenesulfonyl chloride |
| Molecular Weight (g/mol) | 239.12 |
| Formula Weight | 237.96 |