Learn More
3-Chloro-2,4-dimethyl-benzenesulfonyl Chloride, TRC
Product Code. 30362042
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362042 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30362042 Supplier Toronto Research Chemicals Supplier No. TRCC3715755G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chloro-2,4-dimethyl-benzenesulfonyl Chloride
CAS 1783755-80-5
Molecular Formula C8H8Cl2O2S
Quantity 5 g
InChI Formula InChI=1S/C8H8Cl2O2S/c1-5-3-4-7(13(10,11)12)6(2)8(5)9/h3-4H,1-2H3
SMILES Cl[S](=O)(C(C=CC(C)=C1Cl)=C1C)=O
IUPAC Name 3-chloro-2,4-dimethylbenzenesulfonyl chloride
Molecular Weight (g/mol) 239.12
Formula Weight 237.96