Learn More
3-Carbomethoxy-4-hydroxy-Alpha-bromoacetophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Carbomethoxy-4-hydroxy-alpha-bromoacetophenone |
| CAS | 36256-45-8 |
| Molecular Formula | C10H9BrO4 |
| Quantity | 10 g |
| Synonym | 5-(2-Bromoacetyl)-2-hydroxybenzoic Acid Methyl Ester, 5-(Bromoacetyl)salicylic Acid Methyl Ester, Methyl 5-(2-Bromoacetyl)-2-hydroxybenzoate, Methyl 5-(Bromoacetyl)-2-hydroxybenzoate, Methyl 5-(Bromoacetyl)salicylate |
| InChI Formula | InChI=1S/C10H9BrO4/c1-15-10(14)7-4-6(9(13)5-11)2-3-8(7)12/h2-4,12H,5H2,1H3 |
| SMILES | COC(=O)c1cc(ccc1O)C(=O)CBr |
| IUPAC Name | methyl 5-(2-bromoacetyl)-2-hydroxybenzoate |
| Molecular Weight (g/mol) | 273.08 |
| Formula Weight | 271.9684 |