missing translation for 'onlineSavingsMsg'
Learn More

3-Butynyl p-toluenesulfonate, 96%

Product Code. 10125943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10125943 1 g Glass bottle 1g
10771135 5 g Glass bottle 5g
10771325 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10125943 Supplier Thermo Scientific Acros Supplier No. 390780010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23418-85-1 | C11H12O3S | 224.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23418-85-1
Molecular Formula C11H12O3S
Molecular Weight (g/mol) 224.28
MDL Number MFCD00041687
InChI Key STOASOOVVADOKH-UHFFFAOYSA-N
Synonym 3-butynyl p-toluenesulfonate, but-3-yn-1-yl 4-methylbenzenesulfonate, 3-butynyl tosylate, 3-butynyl 4-toluenesulfonate, 3-1-butynyl-4-toluenesulphonate, 3-butynyl 4-methylbenzenesulfonate, p-toluenesulfonic acid 3-butynyl ester, but-3-ynyl tosylate, 4-tosyloxy-1-butyne, acmc-1cgfq
PubChem CID 272020
IUPAC Name but-3-ynyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCC#C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Density 1.0120g/mL
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5220 to 1.524
Quantity 1 g
Specific Gravity 1.012
Formula Weight 224.28
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Butynyl p-toluenesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • 3-Butynyl p-toluenesulfonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.