missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. p-2699962
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
This item is not returnable. View return policy

Product Code. 10293802

Brand: Thermo Scientific Maybridge SEW01928DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 930-96-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00126680
InChI Key BCZHCWCOQDRYGS-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-carboxaldehyde, 3-bromo-2-formylthiophene, 3-bromo-2-thiophenecarbaldehyde, 3-bromo-2-thiophene carboxaldehyde, 3-bromo-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 3-bromo, bromothiophenecarbaldehyde, 3-bromothiophene-2-aldehyde, pubchem5327, acmc-209rjz
PubChem CID 2797079
IUPAC Name 3-bromothiophene-2-carbaldehyde
SMILES BrC1=C(SC=C1)C=O

Specifications

CAS Min % 97
Density 1.81g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 191.05
Percent Purity 97%
Chemical Name or Material 3-Bromothiophene-2-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.