missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyruvic acid, MedChemExpress

Product Code. 16413736
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413736 25 g 25g
16480468 5 g 5g
16451909 1 g 1g
16448008 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16413736 Supplier Medchem Express Supplier No. HY19992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Bromopyruvate (Bromopyruvic acid) is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. 3-Bromopyruvate inhibits cell growth and induces apoptosis through interfering with glycolysis. 3-Bromopyruvate induces autophagy by stimulating ROS formation in breast cancer cells. Antimicrobial activities.

3-Bromopyruvate (Bromopyruvic acid) is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. 3-Bromopyruvate inhibits cell growth and induces apoptosis through interfering with glycolysis. 3-Bromopyruvate induces autophagy by stimulating ROS formation in breast cancer cells. Antimicrobial activities.

Specifications

Chemical Name or Material 3-Bromopyruvic acid
Physical Form Solid
CAS 1113-59-3
Color White
Quantity 25 g
Purity Grade Notes Research
Molecular Formula C3H3BrO3
For Use With (Application) Cancer-Kinase/protease
Synonym Bromopyruvic acid Hexokinase II Inhibitor II, 3-BP
Solubility Information H2O : 250 mg/mL (1497.36 mM; Need ultrasonic)
SMILES O=C(O)C(CBr)=O
Molecular Weight (g/mol) 166.96
Formula Weight 166.96
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.