Learn More
3-Bromopyridine N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromopyridine N-Oxide |
| CAS | 2402-97-3 |
| Molecular Formula | C5 H4 Br N O |
| Quantity | 2 g |
| Synonym | Pyridine, 3-bromo-, 1-oxide, 3-Bromopyridine 1-oxide, 3-Bromopyridine N-oxide, 3-Bromopyridine oxide |
| InChI Formula | InChI=1S/C5H4BrNO/c6-5-2-1-3-7(8)4-5/h1-4H |
| SMILES | [O-][n+]1cccc(Br)c1 |
| IUPAC Name | 3-bromo-1-oxidopyridin-1-ium |
| Molecular Weight (g/mol) | 174 |
| Formula Weight | 172.9476 |