Learn More
3-Bromopropyl-2,5-xylyl Ether, TRC
Product Code. 30343714
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343714 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30343714 Supplier Toronto Research Chemicals Supplier No. TRCB686730250MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Bromopropyl-2,5-xylyl Ethyl
CAS 3245-55-4
Molecular Formula C11 H15 Br O
Quantity 250 mg
Synonym Benzene, 2-(3-bromopropoxy)-1,4-dimethyl-, Ether, 3-bromopropyl 2,5-xylyl (7CI,8CI), 2-(3-Bromopropoxy)-1,4-dimethylbenzene, 1-Bromo-3-(2,5-dimethylphenoxy)propane, 3-(2,5-Dimethylphenoxy)-1-bromopropane, 3-(2,5-Dimethylphenoxy)propyl bromide, 3-(2,5-Xylyloxy)propyl bromide, 3-Bromopropyl 2,5-xylyl ether
InChI Formula InChI=1S/C11H15BrO/c1-9-4-5-10(2)11(8-9)13-7-3-6-12/h4-5,8H,3,6-7H2,1-2H3
SMILES Cc1ccc(C)c(OCCCBr)c1
IUPAC Name 2-(3-bromopropoxy)-1,4-dimethylbenzene
Molecular Weight (g/mol) 243.14
Formula Weight 242.0306