Learn More
3-Bromopropyl-2,5-xylyl Ether, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromopropyl-2,5-xylyl Ethyl |
| CAS | 3245-55-4 |
| Molecular Formula | C11 H15 Br O |
| Quantity | 100 mg |
| Synonym | Benzene, 2-(3-bromopropoxy)-1,4-dimethyl-, Ether, 3-bromopropyl 2,5-xylyl (7CI,8CI), 2-(3-Bromopropoxy)-1,4-dimethylbenzene, 1-Bromo-3-(2,5-dimethylphenoxy)propane, 3-(2,5-Dimethylphenoxy)-1-bromopropane, 3-(2,5-Dimethylphenoxy)propyl bromide, 3-(2,5-Xylyloxy)propyl bromide, 3-Bromopropyl 2,5-xylyl ether |
| InChI Formula | InChI=1S/C11H15BrO/c1-9-4-5-10(2)11(8-9)13-7-3-6-12/h4-5,8H,3,6-7H2,1-2H3 |
| SMILES | Cc1ccc(C)c(OCCCBr)c1 |
| IUPAC Name | 2-(3-bromopropoxy)-1,4-dimethylbenzene |
| Molecular Weight (g/mol) | 243.14 |
| Formula Weight | 242.0306 |