missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobiphenyl, TRC

Product Code. 30005551
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005551 1 g 1g
30137372 5 g 5g
30136326 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005551 Supplier Toronto Research Chemicals Supplier No. TRCB6805551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2113-57-7
Formula Weight 231.9888
InChI Formula InChI=1S/C12H9Br/c13-12-8-4-7-11(9-12)10-5-2-1-3-6-10/h1-9H
IUPAC Name 1-bromo-3-phenylbenzene
Molecular Formula C12 H9 Br
Molecular Weight (g/mol) 233.1
SMILES Brc1cccc(c1)c2ccccc2
Synonym PBB 2 (3-Bromobiphenyl), 1,1'-Biphenyl, 3-bromo-, Biphenyl, 3-bromo- (6CI,7CI,8CI), 3-Bromo-1,1'-biphenyl, 1-Bromo-3-phenylbenzene, 3-Biphenyl bromide, 3-Bromobiphenyl, 3-Xenyl bromide, NSC 407775, PBB 2, [1,1'-Biphenyl]-3-yl bromide, m-Bromobiphenyl
Chemical Name or Material PBB-No. 2

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.