missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzonitrile, 99%

Product Code. p-7021115
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460913 10 g 10g
11470913 50 g 50g
11480913 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460913 Supplier Thermo Scientific Alfa Aesar Supplier No. A13317.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6952-59-6 | C7H4BrN | 182.02 g/mol

3-Bromobenzonitrile is involved in the Suzuki reaction under solvent-free conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzonitrile is involved in the Suzuki reaction under solvent-free conditions.

Solubility
Soluble in water (0.2 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6952-59-6
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD00001796
InChI Key STXAVEHFKAXGOX-UHFFFAOYSA-N
Synonym m-bromobenzonitrile, benzonitrile, 3-bromo, benzonitrile, m-bromo, 3-cyanobromobenzene, 1-bromo-3-cyanobenzene, 3-bromobenzenecarbonitrile, 3-bromo benzonitrile, 3-bromobenzonitride, m-brombenzonitril, 3-brombenzonitrile
PubChem CID 23381
IUPAC Name 3-bromobenzonitrile
SMILES BrC1=CC=CC(=C1)C#N

Specifications

Melting Point 37°C to 41°C
Density 1.562
Boiling Point 224°C to 226°C
Flash Point 113°C (235°F)
Quantity 10 g
UN Number UN3439
Beilstein 1858942
Solubility Information Soluble in water (0.2g/L)
Formula Weight 182.03
Percent Purity 99%
Chemical Name or Material 3-Bromobenzonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.